missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl isocyanate, 97%

Product Code. 15355078 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15355078 5 g 5g
15345078 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15355078 Supplier Thermo Scientific Acros Supplier No. 456490050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 71189-15-6 | C6H5NO2 | 123.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71189-15-6
Molecular Formula C6H5NO2
Molecular Weight (g/mol) 123.11
MDL Number MFCD03180268
InChI Key UIADMYLYGJYUSQ-UHFFFAOYSA-N
Synonym 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl
PubChem CID 4223811
IUPAC Name 2-(isocyanatomethyl)furan
SMILES O=C=NCC1=CC=CO1

Specifications

Melting Point 157.0°C to 158.0°C
Density 1.1480g/mL
Flash Point 54.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.148
Formula Weight 123.11
Percent Purity 97%
Chemical Name or Material Furfuryl isocyanate
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Furfuryl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.