missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl glycidyl ether, 97%

Product Code. 10285721 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10285721 5 g Glass bottle 5g
10357521 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10285721 Supplier Thermo Scientific Acros Supplier No. 348490050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5380-87-0 | C8H10O3 | 154.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5380-87-0
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.17
MDL Number MFCD00013348
InChI Key RUGWIVARLJMKDM-UHFFFAOYNA-N
Synonym furfuryl glycidyl ether, 2-oxiran-2-ylmethoxy methyl furan, 2,3-epoxypropyl-furfurylether, 2-oxiranylmethoxy methyl furan, 2-2,3-epoxypropoxy methyl furan, furan, 2-oxiranylmethoxy methyl, furan, 2-2,3-epoxypropoxy methyl
PubChem CID 94306
IUPAC Name 2-(oxiran-2-ylmethoxymethyl)furan
SMILES C(OCC1=CC=CO1)C1CO1

Specifications

Color Colorless to Orange
Density 1.1220g/mL
Boiling Point 103.0°C to 104.0°C (11.0 mmHg)
Flash Point 102°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4800 to 1.4820
Quantity 5 g
Specific Gravity 1.122
Formula Weight 154.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Furfuryl glycidyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.