missing translation for 'onlineSavingsMsg'
Learn More

Furfuryl acetate, 97%

Product Code. 11448146
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448146 100 g 100g
11438146 25 g 25g
11458146 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448146 Supplier Thermo Scientific Alfa Aesar Supplier No. A15285.22
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 623-17-6 | C7H8O3 | 140.138 g/mol

Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl acetate is a flavoring ingredient. It was used in the synthesis of 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran via Vilsmeier-Haack and Wittig reactions.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 623-17-6
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003251
InChI Key CKOYRRWBOKMNRG-UHFFFAOYSA-N
Synonym furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
PubChem CID 12170
IUPAC Name furan-2-ylmethyl acetate
SMILES CC(=O)OCC1=CC=CO1

Specifications

Density 1.118
Boiling Point 175°C to 177°C
Flash Point 65°C (149°F)
Refractive Index 1.463
Quantity 100 g
Beilstein 116128
Solubility Information Not miscible in water.
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material Furfuryl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.