missing translation for 'onlineSavingsMsg'
Learn More

Furfuraldehyde, Technical, Plastic Coated Container, Fisher Chemical

Product Code. 10599760 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Coated Glass bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10599760 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10599760 Supplier Fisher Chemical Supplier No. F/2080/PB17

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 98-01-1
Melting Point -37°C
Color Amber to Brown
pH 3.5 to 4.5
Boiling Point 159°C
Packaging Plastic Coated Glass bottle
Molecular Formula C5H4O2
MDL Number MFCD00003229
Quantity 2.5 L
Synonym furfural, 2-furaldehyde, 2-furancarboxaldehyde, furaldehyde, 2-formylfuran, furfuraldehyde, fural, 2-furanaldehyde, 2-furancarbonal, 2-furfural
InChI Key HYBBIBNJHNGZAN-UHFFFAOYSA-N
SMILES O=CC1=CC=CO1
IUPAC Name furan-2-carbaldehyde
Molecular Weight (g/mol) 96.09
PubChem CID 7362
ChEBI CHEBI:34768
Formula Weight 96.08g/mol
Vapor Pressure 1mbar at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.