missing translation for 'onlineSavingsMsg'
Learn More

Furfural, Dr. Ehrenstorfer

Product Code. 30324844
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324844 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324844 Supplier Dr. Ehrenstorfer Supplier No. DREC13972100100MG
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furfural
CAS 98-01-1
Analyte or Component Names Furfural
Shipping Condition +20°C
Molecular Formula C5 H4 O2
Certifications/Compliance ISO 17034
Synonym Furane-2-carbaldehyde, Furfural, 2-Furfurol, 2-Furaldehyde, Ascorbic Acid Impurity A, Ethanol Impurity N, Sodium Ascorbate Impurity A
InChI Formula InChI=1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H
SMILES O=Cc1occc1
IUPAC Name furan-2-carbaldehyde
Molecular Weight (g/mol) 96.08
Quantity 100 mg
Formula Weight 96.0211 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.