missing translation for 'onlineSavingsMsg'
Learn More

Furfural 2,4-Dinitrophenylhydrazone, TRC

Product Code. 30400440
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400440 250 mg
30404112 50 mg
30403860 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400440 Supplier Toronto Research Chemicals Supplier No. TRCF862380250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2074-02-4
Molecular Formula C11 H8 N4 O5
Molecular Weight (g/mol) 276.2
Synonym 2-Furaldehyde, (2,4-dinitrophenyl)hydrazone (6CI,7CI,8CI), 2-Furancarboxaldehyde, (2,4-dinitrophenyl)hydrazone (9CI), Furfural 2,4-dinitrophenylhydrazone
IUPAC Name N-(furan-2-ylmethylideneamino)-2,4-dinitroaniline
SMILES [O-][N+](=O)c1ccc(NN=Cc2occc2)c(c1)[N+](=O)[O-]

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C11H8N4O5/c16-14(17)8-3-4-10(11(6-8)15(18)19)13-12-7-9-2-1-5-20-9/h1-7,13H
Formula Weight 276.0495
Chemical Name or Material Furfural-2,4-dinitrophenylhydrazone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.