missing translation for 'onlineSavingsMsg'
Learn More

Furaprofen, MedChemExpress

Product Code. 16452448
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452448 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16452448 Supplier Medchem Express Supplier No. HYU00213
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Furaprofen (R803) is an effective HCV replication inhibitor. Furaprofen (R803) is substantially more potent against genotype 1a and 1b replicons (EC50, ∼30 nM) than against the genotype 2a replicon (EC50, ∼1,000 nM).

Furaprofen (R803) is an effective HCV replication inhibitor. Furaprofen (R803) is substantially more potent against genotype 1a and 1b replicons (EC50, ∼30 nM) than against the genotype 2a replicon (EC50, ∼1,000 nM).

Specifications

Chemical Name or Material Furaprofen
CAS 67700-30-5
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H14O3
Quantity 5 mg
Synonym R803
Solubility Information DMSO : 250 mg/mL (938.83 mM; Need ultrasonic)
SMILES OC(C(C1=CC=CC2=C1OC=C2C3=CC=CC=C3)C)=O
Molecular Weight (g/mol) 266.29
Formula Weight 266.29
Percent Purity 99.95%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.