missing translation for 'onlineSavingsMsg'
Learn More

Furan (98%), TRC

Product Code. 30395449
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30395449 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395449 Supplier Toronto Research Chemicals Supplier No. TRCF863650100MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furan
CAS 110-00-9
Purity Grade Notes GC
Molecular Formula C4 H4 O
Quantity 100 mg
Synonym Divinylene oxide, Furfuran, Oxacyclopentadiene, Oxole, Tetrole
InChI Formula InChI=1S/C4H4O/c1-2-4-5-3-1/h1-4H
SMILES o1cccc1
IUPAC Name furan
Molecular Weight (g/mol) 68.07
Formula Weight 68.0262
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.