missing translation for 'onlineSavingsMsg'
Learn More

Furan-3-boronic acid, 97%

Product Code. 11349815
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349815 1 g 1g
11359815 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349815 Supplier Thermo Scientific Alfa Aesar Supplier No. L19834.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55552-70-0 | C4H5BO3 | 111.89 g/mol

Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furan-3-boronic acid use furan-3-boronic acid as our coupling partner in many reactions. A chitosan uricase (UOx)-poly (furan-3-boronic acid)(PFBA)-Pd nanoparticles (PdNPs)/plated Pd (Pd plate)/multiwalled carbon nanotubes (MWCNTs)/Au electrode was prepared for fabricating an amperometric biosensor and a biofuel cell (BFC) of uric acid.

Solubility
Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55552-70-0
Molecular Formula C4H5BO3
Molecular Weight (g/mol) 111.89
MDL Number MFCD01319007
InChI Key CYEFKCRAAGLNHW-UHFFFAOYSA-N
Synonym furan-3-boronic acid, 3-furanboronic acid, 3-furylboronic acid, furan-3-yl boronic acid, 3-furanylboronic acid, furan-3-ylboranediol, 3-furan boronic acid, furan-3-yl-3-boronic acid, furan-3-yl-boronic acid, boronic acid, 3-furanyl
PubChem CID 2734358
IUPAC Name (furan-3-yl)boronic acid
SMILES OB(O)C1=COC=C1

Specifications

Melting Point 139°C to 144°C
Quantity 1 g
Beilstein 1635653
Solubility Information Soluble in methanol.
Formula Weight 111.89
Percent Purity 97%
Chemical Name or Material Furan-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.