missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-ylmethyl-phenethyl-amine, TRC

Product Code. 30395955
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395955 500 mg
30400663 100 mg
30402460 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395955 Supplier Toronto Research Chemicals Supplier No. TRCF864208500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4439-55-8
Molecular Formula C13H15NO
Molecular Weight (g/mol) 201.26
Synonym N-(2-Phenylethyl)-2-furanmethanamine, (Furan-2-ylmethyl)(2-phenylethyl)amine, [(Furan-2-yl)methyl](2-phenylethyl)amine, N-phenethyl-Furfurylamine
SMILES C(Cc1ccccc1)NCc2occc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H15NO/c1-2-5-12(6-3-1)8-9-14-11-13-7-4-10-15-13/h1-7,10,14H,8-9,11H2
Formula Weight 201.1154
Chemical Name or Material Furan-2-ylmethyl-phenethyl-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.