missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furan-2-carbaldehyde (Furfural), Mikromol™
Description
Furan-2-carbaldehyde (Furfural)
Specifications
Specifications
| Chemical Name or Material | Furfural |
| CAS | 98-01-1 |
| Analyte or Component Names | Furfural |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H4 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Furane-2-carbaldehyde, Furfural, 2-Furfurol, 2-Furaldehyde, Ascorbic Acid Impurity A, Ethanol Impurity N, Sodium Ascorbate Impurity A |
| InChI Formula | InChI=1S/C5H4O2/c6-4-5-2-1-3-7-5/h1-4H |
| SMILES | O=Cc1occc1 |
| IUPAC Name | furan-2-carbaldehyde |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Individuate un'opportunità di miglioramento?