missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-boronic acid, TRC

Product Code. 30391911
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30391911 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30391911 Supplier Toronto Research Chemicals Supplier No. TRCF864095500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13331-23-2
Molecular Formula C4H5BO3
Quantity 500 mg
InChI Formula InChI=1S/C4H5BO3/c6-5(7)4-2-1-3-8-4/h1-3,6-7H
SMILES OB(O)c1occc1
Molecular Weight (g/mol) 111.89
Formula Weight 112.0332
Chemical Name or Material Furan-2-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.