missing translation for 'onlineSavingsMsg'
Learn More

Furan-2-boronic acid, 97%

Product Code. p-7027245
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11496686

Brand: Thermo Scientific Alfa Aesar B23842.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13331-23-2 | C4H5BO3 | 111.891 g/mol

It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in Suzuki reaction for generalized route for the synthesis of β-furyl-α,β-unsaturated aldehydes.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13331-23-2
Molecular Formula C4H5BO3
Molecular Weight (g/mol) 111.891
MDL Number MFCD00799544
InChI Key PZJSZBJLOWMDRG-UHFFFAOYSA-N
Synonym 2-furanboronic acid, furan-2-boronic acid, 2-furylboronic acid, 2-furanylboronic acid, furan-2-ylboranediol, furan 2-boronic acid, 2-furan boronic acid, furan-2-yl boronic acid, boronic acid, furanyl, 2furylboronic acid
PubChem CID 2734357
IUPAC Name furan-2-ylboronic acid
SMILES B(C1=CC=CO1)(O)O

Specifications

Melting Point ∼120°C (decomposition)
Quantity 1 g
Beilstein 112370
Sensitivity Air sensitive
Solubility Information Slightly soluble in water.
Formula Weight 111.89
Percent Purity 97%
Chemical Name or Material Furan-2-boronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.