missing translation for 'onlineSavingsMsg'
Learn More

Furan-2,5-dicarboxylic acid, 98%

Product Code. p-7030435
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11452120

Brand: Thermo Scientific Alfa Aesar H28718.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3238-40-2 | C6H4O5 | 156.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3238-40-2
Molecular Formula C6H4O5
Molecular Weight (g/mol) 156.09
MDL Number MFCD00016582
InChI Key CHTHALBTIRVDBM-UHFFFAOYSA-N
Synonym 2,5-furandicarboxylic acid, dehydromucic acid, furan-2,5-dicarbonsaeure, 2,5-furandicarboxylicacid, 2,5-furandicarboxylate, furane-alpha,alpha'-dicarboxylic acid, furan 2,5-dicarboxylic acid, furan-2,5-dicarboxylate, furane-.alpha.,.alpha.'-dicarboxylic acid, 2,5 furan dicarboxylic acid
PubChem CID 76720
ChEBI CHEBI:84212
IUPAC Name furan-2,5-dicarboxylic acid
SMILES OC(=O)C1=CC=C(O1)C(O)=O

Specifications

Melting Point >310°C
Quantity 1 g
Formula Weight 156.09
Percent Purity 98%
Chemical Name or Material Furan-2,5-dicarboxylic acid
compliance-icons
Product Identifier
  • Furan-2,5-dicarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,5-Furandicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.