missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Furaltadone hydrochloride, MedChemExpress
Description
Furaltadone hydrochloride, a nitrofuran drug, has the potential for the study in infections of chickens with salmonella enteritidis. Furaltadone is inhibitory and bactericidal in vitro for staphylococci .
Specifications
Specifications
| Chemical Name or Material | Furaltadone hydrochloride |
| CAS | 3759-92-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C13H17ClN4O6 |
| Quantity | 100 mg |
| Synonym | Altafur hydrochloride |
| Solubility Information | DMSO : 16.67 mg/mL (46.21 mM; Need ultrasonic) |
| SMILES | O=C1OC(CN2CCOCC2)CN1/N=C/C3=CC=C([N+]([O-])=O)O3.[H]Cl |
| Molecular Weight (g/mol) | 360.75 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?