missing translation for 'onlineSavingsMsg'
Learn More

Furagin, MedChemExpress

Product Code. 16451979
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16451979

Brand: Medchem Express HY77036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Furagin, nitrofurantoin analog, is an anti-bacterial agent.

Furagin, nitrofurantoin analog, is an anti-bacterial agent.

Specifications

Chemical Name or Material Furagin
Physical Form Solid
CAS 1672-88-4
Color Yellow
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C10H8N4O5
For Use With (Application) COVID-19-immunoregulation
Synonym Furazidine Furazidin
Solubility Information DMSO : 25 mg/mL (94.63 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1NC(CN1/N=C/C=C/C2=CC=C(O2)[N+]([O-])=O)=O
Molecular Weight (g/mol) 264.19
Formula Weight 264.19
Percent Purity 98.6%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.