missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUOM Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580646
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human FUOM/fucose mutarotase/FucM.
Involved in the interconversion between alpha- and beta-L-fucoses. L-Fucose (6-deoxy-L-galactose) exists as alpha-L-fucose (29.5%) and beta-L-fucose (70.5%), the beta-form is metabolized through the salvage pathway. GDP-L-fucose formed either by the de novo or salvage pathways is transported into the endoplasmic reticulum, where it serves as a substrate for N- and O-glycosylations by fucosyltransferases. Fucosylated structures expressed on cell surfaces or secreted in biological fluids are believed to play a critical role in cell-cell adhesion and recognition processes.
Specifications
| FUOM | |
| Polyclonal | |
| Unconjugated | |
| FUOM | |
| C10orf125; FucM; Fucose mutarotase; FUCU; FUOM; protein fucU homolog | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 282969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| A2VDF0 | |
| FUOM | |
| Recombinant Human FUOM/fucose mutarotase/FucM Protein (Met1-Leu154). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction