missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ Funnel for Barrel ARNOLD, ASTM 63

Product Code. 31454258 Shop All SCAT Europe Products
Change view
Click to view available options
Closure Type:
Hinged Lid
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Type
31454258 Hinged Lid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31454258 Supplier SCAT™ Supplier No. 317652
  • ONLINE EXCLUSIVE PRICE
    £211.00
    £161.52 / Pack of 1
    Save £49.48
    23% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Robust SCAT™ 'ARNOLD' Safety Funnels with adapters and mechanical level controls for use on various drums with different thread types. Also available in electrically dissipative variants for the safe disposal of explosive liquid waste.

  • Safety funnels based on the SCAT™ 'ARNOLD' funnel series
  • Ready-to-use sets consisting of safety funnels and barrel adapters
  • Made of high-quality plastics for durable and long-lasting use
  • Adapters can be combined with all GL45 safety funnels of the 'ARNOLD' and 'MARCO' series
  • Available for all common thread types and sizes
  • Also available in electrically conductive material for hazardous areas

Specifications

Capacity (Metric) 650 mL
Closure Type Hinged Lid
Color Black
Height (Metric) 210 mm
Includes Lid, Splash Guard, Removable Sieve, Grounding Cable, Ball Valve
For Use With (Equipment) SCAT ARNOLD Funnel
Material HDPE-EC
Product Type Safety Funnel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.