missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fungal beta glucan Recombinant Rabbit Monoclonal Antibody (8201)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA533305
This item is not returnable.
View return policy
Description
Reactive with Candida (5 species), Aspergillus (4 species), Penicillium notatum, Saccharomyces cerevisiae, Pichia pastoris, and Mucor hiemalis. Others not tested. Extraction of specimen with 0.4M NaOH required for optimum sensitivity.
Beta-glucans are naturally occurring glucose polymers that are present in abundance in fungi cell walls.
Specifications
| Fungal beta glucan | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.5 | |
| Protein A | |
| RUO | |
| Fungi | |
| Antibody | |
| IgG1 |
| ELISA, Immunocytochemistry | |
| 8201 | |
| Unconjugated | |
| Rabbit | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction