missing translation for 'onlineSavingsMsg'
Learn More

Fumonisins B1 and B2 Mixture 50 μg/mL in Acetonitrile/Water, Dr. Ehrenstorfer

Product Code. 30317928
Change view
Click to view available options
Quantity:
1 mL
5 mL
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317928 1 mL 1mL
30328239 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30317928 Supplier Dr. Ehrenstorfer Supplier No. DREA30000003WL
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fumonisin B1∣Fumonisin B2
CAS 116355-83-0, 116355-84-1
Analyte or Component Names Fumonisin B1∣Fumonisin B2
Shipping Condition Room Temperature
Certifications/Compliance ISO 9001
Solution Type Mixture
Solvent or Matrix Acetonitrile/Water
Industry Type Environmental Testing
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.