missing translation for 'onlineSavingsMsg'
Learn More

Fumonisin B2, TRC

Product Code. 18031040
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
5 mg
Unit Size:
0.5mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18031040 0.5 mg 0.5mg
18081930 5 mg 5mg
18091240 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18031040 Supplier Toronto Research Chemicals Supplier No. TRCF8627550.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116355-84-1
Formula Weight 705.3936
InChI Formula InChI=1S/C34H59NO14/c1-5-6-12-21(3)32(49-31(43)18-24(34(46)47)16-29(40)41)27(48-30(42)17-23(33(44)45)15-28(38)39)14-20(2)11-9-7-8-10-13-25(36)19-26(37)22(4)35/h20-27,32,36-37H,5-19,35H2,1-4H3,(H,38,39)(H,40,41)(H,44,45)(H,46,47)/t20-,21+,22-,23+,24+,25+,26-,27-,32+/m0/s1
IUPAC Name (2R)-2-[2-[(5R,6R,7S,9S,16R,18S,19S)-19-amino-6-[(3R)-3,4-dicarboxybutanoyl]oxy-16,18-dihydroxy-5,9-dimethylicosan-7-yl]oxy-2-oxoethyl]butanedioic acid
Molecular Formula C34 H59 N O14
Molecular Weight (g/mol) 705.83
Percent Purity >95%
SMILES CCCC[C@@H](C)[C@@H](OC(=O)C[C@@H](CC(=O)O)C(=O)O)[C@H](C[C@@H](C)CCCCCC[C@@H](O)C[C@H](O)[C@H](C)N)OC(=O)C[C@@H](CC(=O)O)C(=O)O
Synonym 1,2,3-Propanetricarboxylic acid, 1,1'-[1-(12-amino-9,11-dihydroxy-2-methyltridecyl)-2-(1-methylpentyl)-1,2-ethanediyl] ester, [2S-[1[1R*(S*),2S*(S*),2(S*)],2R*,9S*,11R*,12R*]]-, Fumonisin B2
Chemical Name or Material Fumonisin B2
Grade HPLC
Show More Show Less

Specifications

Quantity 0.5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.