missing translation for 'onlineSavingsMsg'
Learn More

Fumonisin B1, 96%

Product Code. p-3778694 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 10741361

Brand: Thermo Scientific Acros 328250010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Fumonisin, CAS # 116355-83-0, also known as Macrofusine, is a cell permeable mycotoxin that is an inhibitor of sphingosine N-acetyl transferase. | CAS: 116355-83-0 | C34H59NO15 | 721.8 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fumonisin is a fungal metabolite that is a cell permeable mycotoxin that can halt sphingolipid biosynthesis by inhibiting sphingosine N-acetyl transferase

Applications

• It can potently inhibit sphingosine N-acyltransferase (ceramide synthase), causing an accumulation of sphingoid bases

• It is also a protein serine and threonine phosphatase inhibitor

• It can selectively inhibit sphingomyelin biosynthesis in neuronal cells

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116355-83-0
Molecular Formula C34H59NO15
Molecular Weight (g/mol) 721.8
MDL Number MFCD00133349
InChI Key UVBUBMSSQKOIBE-UWCNXKJPSA-N
Synonym macrofusine, fumonisin b1, 2r,2'r-1,2,3-propanetricarboxylic acid 1,1'-1s,2r-1-2s,4r,9r,11s,12s-12-amino-4,9,11-trihydroxy-2-methyltridecyl-2-1r-1-methylpentyl-1,2-ethanediyl ester, 2s-2-2-5r,6r,7s,9s,11r,16r,18s,19s-19-amino-6-3r-3,4-dicarboxybutanoyl oxy-11,16,18-trihydroxy-5,9-dimethylicosan-7-yl oxy-2-oxoethyl butanedioic acid
PubChem CID 16760030
IUPAC Name (2S)-2-[2-[(5R,6R,7S,9S,11R,16R,18S,19S)-19-amino-6-[(3R)-3,4-dicarboxybutanoyl]oxy-11,16,18-trihydroxy-5,9-dimethylicosan-7-yl]oxy-2-oxoethyl]butanedioic acid
SMILES CCCCC(C)C(C(CC(C)CC(CCCCC(CC(C(C)N)O)O)O)OC(=O)CC(CC(=O)O)C(=O)O)OC(=O)CC(CC(=O)O)C(=O)O

Specifications

Color Tan to White
Elemental Analysis 55.00 to 58.00%C, 1.60 to 2.20%N
Assay Percent Range 95% min. (HPLC)
Quantity 1 mg
Merck Index 15, 4318
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,acetonitrile
Formula Weight 721.8
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Fumonisin B1
compliance-icons
Product Identifier
  • Fumonisin B1
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.