missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Fumonisin B1

Product Code. 17130262
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17130262 1 mg 1mg
17140262 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17130262 Supplier STEMCELL Technologies Supplier No. 73682
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibitor of sphingolipid synthesis and protein serine/threonine phosphatases

  • Fumonisin B1 is a mycotoxin produced by Fusarium moniliforme that has been shown to potently inhibit sphingosine N-acyltransferase (ceramide synthase; Wang et al.), thereby disrupting the synthesis of sphingolipids, a key component of plasma membranes (IC50 = 0.1 μM)
  • Fumonisin B1 also inhibits protein serine/threonine phosphatases (PPs; PP1, PP2A, PP2B, PP2C, and PP5/T/K/H) with IC50 values of 80 - 3000 μM
  • PP5 is the most sensitive with an IC50 of 80 μM (Fukuda et al.) Fumonisin B1, together with aflatoxin B1, increases reactive oxygen species levels and oxidative damage in rat spleen cells (Mary et al.)

Specifications

CAS 116355-83-0
Quantity 1 mg
Molecular Formula C34H59NO15
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.