missing translation for 'onlineSavingsMsg'
Learn More

Fumaryl chloride, 95%

Product Code. 11491908
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11491908 25 g 25g
11401918 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11491908 Supplier Thermo Scientific Alfa Aesar Supplier No. A14540.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-63-4 | C4H2Cl2O2 | 152.958 g/mol

Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fumaryl chloride was used in the preparation of high molecular weight poly(propylene fumarate). The photodissociation dynamics of fumaryl chloride was studied. It is used in agrochemicals and pharmaceuticals manufacturing. Also used as an intermediate for dyes, textile auxiliaries and peroxide compounds. It is an acid halide building block.

Solubility
It reacts with water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, active metals and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-63-4
Molecular Formula C4H2Cl2O2
Molecular Weight (g/mol) 152.958
MDL Number MFCD00000733
InChI Key ZLYYJUJDFKGVKB-OWOJBTEDSA-N
Synonym fumaryl chloride, fumaroyl dichloride, fumaroyl chloride, fumarylchlorid, chlorure de fumaryle, fumarylchlorid czech, 2-butenedioyl dichloride, e, 2-butenedioyl dichloride, 2e, dichlorid kyseliny fumarove, fumaryl dichloride
PubChem CID 5325504
IUPAC Name (E)-but-2-enedioyl dichloride
SMILES C(=CC(=O)Cl)C(=O)Cl

Specifications

Melting Point -2°C
Density 1.413
Boiling Point 159°C to 161°C
Flash Point 73°C (163°F)
Odor Acrid
Refractive Index 1.5
Quantity 25 g
UN Number UN1780
Beilstein 1721342
Sensitivity Moisture sensitive
Solubility Information It reacts with water.
Formula Weight 152.97
Percent Purity 95%
Chemical Name or Material Fumaryl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.