missing translation for 'onlineSavingsMsg'
Learn More

Fumaronitrile, 95%

Product Code. 10295711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295711 5 g Glass bottle 5g
10397891 25 g Glass bottle 25g
10657294 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295711 Supplier Thermo Scientific Acros Supplier No. 157400050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 764-42-1 | C4H2N2 | 78.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 764-42-1
Molecular Formula C4H2N2
Molecular Weight (g/mol) 78.06
MDL Number MFCD00001928
InChI Key KYPOHTVBFVELTG-OWOJBTEDSA-N
Synonym fumaronitrile, fumarodinitrile, furmaronitrile, fumaric nitrile, 2-butenedinitrile, e, 2-butenedinitrile, 2e, trans-1,2-dicyanoethylene, trans-1,2-dicyanoethene, 2e-but-2-enedinitrile, e-1,2-dicyanoethylene
PubChem CID 637930
IUPAC Name (E)-but-2-enedinitrile
SMILES C(=CC#N)C#N

Specifications

Melting Point 95.0°C to 98.0°C
Color White
Boiling Point 186.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula NCCH=CHCN
Quantity 5 g
Beilstein 02, I, 302
Formula Weight 78.06
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Fumaronitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.