missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid tert-Butyl Ester (>90%), TRC

Product Code. 30400619
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400619 5 g
30397899 500 mg
30406516 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400619 Supplier Toronto Research Chemicals Supplier No. TRCF5003955G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135355-96-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
Synonym (2E)-2-Butenedioic Acid Mono(1,1-dimethylethyl) Ester, (E)-4-tert-Butoxy-4-oxobut-2-enoic Acid
SMILES CC(C)(C)OC(=O)\C=C\C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C8H12O4/c1-8(2,3)12-7(11)5-4-6(9)10/h4-5H,1-3H3,(H,9,10)/b5-4+
Formula Weight 172.0736
Chemical Name or Material Fumaric Acid tert-Butyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.