missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid tert-Butyl Ester (>90%), TRC

Product Code. 30397899
Change view
Click to view available options
Quantity:
500 mg
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397899 500 mg
30400619 5 g
30406516 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397899 Supplier Toronto Research Chemicals Supplier No. TRCF500395500MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 135355-96-3
Molecular Formula C8H12O4
Molecular Weight (g/mol) 172.18
Synonym (2E)-2-Butenedioic Acid Mono(1,1-dimethylethyl) Ester, (E)-4-tert-Butoxy-4-oxobut-2-enoic Acid
SMILES CC(C)(C)OC(=O)\C=C\C(=O)O

Specifikationer

Quantity 500 mg
InChI Formula InChI=1S/C8H12O4/c1-8(2,3)12-7(11)5-4-6(9)10/h4-5H,1-3H3,(H,9,10)/b5-4+
Formula Weight 172.0736
Chemical Name or Material Fumaric Acid tert-Butyl Ester
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.