missing translation for 'onlineSavingsMsg'
Learn More

Fumaric Acid Monoethyl Ester, TRC

Product Code. 30394886
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394886 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394886 Supplier Toronto Research Chemicals Supplier No. TRCF5003851G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2459-05-4
Molecular Formula C6 H8 O4
Quantity 1 g
Synonym (2E)-2-Butenedioic acid 1-ethyl ester, 2-Butenedioic acid (2E)-, monoethyl ester (9CI), 2-Butenedioic acid (E)-, monoethyl ester, Fumaric acid, ethyl ester (6CI,7CI), Fumaric acid, monoethyl ester (8CI), (E)-4-Ethoxy-4-oxo-2-butenoic acid, Ethyl hydrogen fumarate, Monoethyl fumarate, Monoethyl fumerate, Monoethyl trans-2-butenedioate
InChI Formula InChI=1S/C6H8O4/c1-2-10-6(9)4-3-5(7)8/h3-4H,2H2,1H3,(H,7,8)/b4-3+
SMILES CCOC(=O)\C=C\C(=O)O
IUPAC Name (E)-4-ethoxy-4-oxobut-2-enoic acid
Molecular Weight (g/mol) 144.13
Formula Weight 144.0423
Chemical Name or Material Monoethyl Fumarate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.