missing translation for 'onlineSavingsMsg'
Learn More

FumARic Acid, Extra Pure, SLR, Fisher Chemical

Product Code. 10775521 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10775521 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10775521 Supplier Fisher Chemical Supplier No. F/2070/53

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 110-17-8
Melting Point 295°C
Color White
Packaging Plastic powder jar
Molecular Formula C4H4O4
MDL Number MFCD00002700
Quantity 500 g
Synonym fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES OC(=O)\C=C\C(O)=O
IUPAC Name (E)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
PubChem CID 444972
ChEBI CHEBI:18012
Formula Weight 116.07g/mol
Physical Form Powder
Show More Show Less
compliance-icons
Product Identifier
  • Fumaric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.