missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumaric Acid-d2, TRC
Specifications
Specifications
| Chemical Name or Material | Fumaric Acid-d2 |
| CAS | 24461-32-3 |
| Molecular Formula | C4 D2 H2 O4 |
| Quantity | 100 mg |
| Synonym | (2E)-2-Butenedioic Acid-d2, (2E)-But-2-enedioic Acid-d2, (E)-2-Butenedioic Acid-d2, 2-(E)-Butenedioic Acid-d2, (E)-Butenedioic Acid-d2, Allomaleic Acid-d2, Bakeshure 451-d2, Bakeshure 470-d2, Boletic Acid-d2, FC 33-d2, Lichenic Acid-d2, NSC 2752-d2, Petrom-d2, trans-1,2-Ethylenedicarboxylic Acid-d2, trans-2-Butenedioic Acid-d2, trans-Butenedioic Acid-d2 |
| InChI Formula | InChI=1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1+/i1D,2D |
| SMILES | [2H]\C(=C(\[2H])/C(=O)O)\C(=O)O |
| IUPAC Name | (E)-2,3-dideuteriobut-2-enedioic acid |
| Molecular Weight (g/mol) | 118.08 |
| Formula Weight | 118.0235 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?