missing translation for 'onlineSavingsMsg'
Learn More

Fumaric acid, 99+%

Product Code. p-3598377 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
100g
10kg
1kg
2.5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10286233 100 g Plastic bottle 100g
10274720 1 kg Plastic bottle 1kg
10744801 2.5 kg Plastic bottle 2.5kg
10423025 10 kg Plastic drum 10kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10286233 Supplier Thermo Scientific Acros Supplier No. 119751000
Add to basket
This item is not returnable. View return policy

CAS: 110-17-8 | C4H4O4 | 116.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-17-8
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00002700
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
PubChem CID 444972
ChEBI CHEBI:18012
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 295.0°C to 300.0°C
Color White
Density 1.6200g/mL
Flash Point 230°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula HO2CCH=CHCO2H
Quantity 100 g
Beilstein 02, IV, 2202
Fieser 05,319
Merck Index 15, 4316
Specific Gravity 1.62
Solubility Information Solubility in water: 6.3g/l (25°C). Other solubilities: 0.72g/100g ether (25°C)- 1.72g/100g acetone,(30°C)- 5.76g/100 g 95% alcohol (30°C),practically insoluble in chloroform,carbon tetra-,chloride and benzene
IUPAC Name (2E)-but-2-enedioic acid
Formula Weight 116.07
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material Fumaric acid
Show More Show Less
compliance-icons
Product Identifier
  • Fumaric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.