missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fumarase Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582967
This item is not returnable.
View return policy
Description
Immunogen sequence: HFPLVVWQTG SGTQTNMNVN EVISNRAIEM LGGELGSKIP VHPNDHVNKS QSSNDTFPTA MHIAAAIEVH EVLLPGLQKL HDALDAKSKE FAQIIK Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 95%.
The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy.
Specifications
| Fumarase | |
| Polyclonal | |
| Unconjugated | |
| FH | |
| EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2271 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P07954 | |
| FH | |
| Recombinant protein corresponding to Human FH. Recombinant protein control fragment (Product #RP-94049). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto