missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fumarase Monoclonal Antibody (OTI1F10)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524989
This item is not returnable.
View return policy
Descrizione
Fumarase Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy.
Specifica
| Fumarase | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P07954, P14408, P97807 | |
| FH, Fh1 | |
| Full length human recombinant protein of FH produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1F10 | |
| Unconjugated | |
| FH | |
| EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 14193, 14194, 2271, 24368, 480092 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto