missing translation for 'onlineSavingsMsg'
Learn More

Fumaraldehyde bis(dimethylacetal), 90%, Thermo Scientific™

Product Code. 10310814 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10310814 5 g Glass bottle 5g
10425665 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10310814 Supplier Thermo Scientific Acros Supplier No. 388530050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6068-62-8
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00075507
InChI Key ZFGVCDSFRAMNMT-AATRIKPKSA-N
Synonym fumaraldehyde bis dimethyl acetal, e-1,1,4,4-tetramethoxybut-2-ene, fumaraldehyde bis dimethylacetal, 2-butenedial bis dimethyl acetal, 1,1,4,4-tetramethoxy-trans-2-butene, trans-1,1,4,4-tetramethoxy-2-butene, 1,1,4,4-tetramethoxy-but-2-ene
PubChem CID 5862719
IUPAC Name (E)-1,1,4,4-tetramethoxybut-2-ene
SMILES COC(C=CC(OC)OC)OC

Specifications

Color Colorless to Yellow
Density 1.0100g/mL
Boiling Point 100°C to 103°C (15.0 mmHg)
Flash Point 80°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2CHCH=CHCH(OCH3)2
Quantity 5 g
Specific Gravity 1.01
Formula Weight 176.21
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Fumaraldehyde bis(dimethylacetal)
Show More Show Less
compliance-icons
Product Identifier
  • Fumaraldehyde bis(dimethylacetal)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.