missing translation for 'onlineSavingsMsg'
Learn More

Fumagillin, MedChemExpress

Product Code. 16453796
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16453796

Brand: Medchem Express HYB0751

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fumagillin(NSC9168) is an antimicrobial compound first isolated in 1949 from the fungus Aspergillus fumigatu. Fumagillin can inhibits HIV‐1 infection through the inhibition of HIV-1 viral protein R (Vpr) activity.

Fumagillin(NSC9168) is an antimicrobial compound first isolated in 1949 from the fungus Aspergillus fumigatu. Fumagillin can inhibits HIV‐1 infection through the inhibition of HIV-1 viral protein R (Vpr) activity.

Specifications

Chemical Name or Material Fumagillin
Physical Form Solid
CAS 23110-15-8
Color Off-White
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C26H34O7
For Use With (Application) COVID-19-anti-virus
Synonym Amebacilin NSC9168
Solubility Information DMSO : 50 mg/mL (109.04 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES C[C@]1([C@@H](C/C=C(C)/C)O1)[C@]([C@@H]2OC)([H])[C@]3(CC[C@H]2OC(/C=C/C=C/C=C/C=C/C(O)=O)=O)CO3
Molecular Weight (g/mol) 458.54
Formula Weight 458.54
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.