Learn More
Thermo Scientific Alfa Aesar Fullerene powder, mixed hydrogenated, typically 77% C{60}H{x}, 22% C{70}H{y}
Brand: Thermo Scientific Alfa Aesar 040967.02
Fullerene powder, mixed hydrogenated, typically 77% C{60}H{x}, 22% C{70}H{y}
Code : NEW
Additional Details : Weight : 0.00050kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III