missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, hydroxylated, C{60}(OH)∣n,, Thermo Scientific Chemicals

Product Code. 11349557
Click to view available options
Quantity:
25 mg
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99685-96-8
Molecular Formula C60
Molecular Weight (g/mol) 720.66
MDL Number MFCD06202029
InChI Key XMWRBQBLMFGWIX-UHFFFAOYSA-N
Synonym fullerene, buckminsterfullerene, buckyball, fullerene 60, 60 fullerene, footballene, fullerene-c60, 5,6 fullerene-c60-ih
PubChem CID 123591
ChEBI CHEBI:33128
IUPAC Name (C\{60}-I\{h})[5,6]fullerene
SMILES C12=C3C4=C5C1=C1C6=C7C2=C2C8=C3C3=C9C4=C4C%10=C5C5=C1C1=C6C6=C%11C7=C2C2=C7C8=C3C3=C8C9=C4C4=C9C%10=C5C5=C1C1=C6C6=C%11C2=C2C7=C3C3=C8C4=C4C9=C5C1=C1C6=C2C3=C41

Specifications

Quantity 25 mg
UN Number UN1325
Physical Form Powder
Chemical Name or Material Fullerene powder, Hydroxylated, C60(OH)n

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'