Learn More
Fullerene powder, 99% C{6}{0}
Description
It is used in optical computing, optical memories, optical signal processing and controlling applications respects, C60 and its derivatives can be widely used in photovoltaic applications, magnetic resonance imaging, anti-HIV drugs, anti-cancer drugs, chemotherapy drugs, cosmetic additives, research and other areas.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| Density | 1.7 g/mL |
| Boiling Point | 500°C to 600°C (sublimation) |
| Quantity | 1 g |
| UN Number | UN1325 |
| Solubility Information | Soluble in organic solvents. Insoluble in water. |
| Percent Purity | 99% |
| Physical Form | Powder |
| Assay | C60 |
| Chemical Name or Material | Fullerene powder |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.