Learn More
Fullerene powder, 99.9+% C{60}, Thermo Scientific Chemicals
Description
Fullerene powder is useful for biomedical applications such as the design of high-performance MRI contrast agents, x-ray imaging contrast agents and photodynamic therapy. It is also essential for single-walled carbon nanotubes with narrow diameter-distribution. It is used as light-activated antimicrobial agent. It also finds application in the field of nanotechnology, heat resistance and superconductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Black |
Density | 1.7g/mL |
Boiling Point | 500°C to 600°C (sublimation) |
Assay Percent Range | >99.9% C60 |
Quantity | 5 g |
UN Number | UN1325 |
Percent Purity | ≥99.9% |
Physical Form | Powder |
Assay | C60 |
Chemical Name or Material | Fullerene powder |
RUO – Research Use Only