missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Fullerene powder, 99.5% C{60}

Brand:  Alfa Aesar™ 039722.03

Code : NEW

Additional Details : CAS Number : 99685-96-8 Weight : 0.00100kg Transport : UN number : 1325 Chem class : 1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 11394677

  • £126.50 / 1g


Estimated Shipment date
from Supplier 24-01-2018
Add to basket

Specifications

Chemical Identifiers

Fullerene powder
MFCD00151408
Buckminsterfullerene, C60
99685-96-8
UN1325

Specifications

Fullerene powder
Powder
C60
UN1325
1.7g/mL
Black/brown
99685-96-8
99.5%
MFCD00151408
Buckminsterfullerene, C60
500° to 600°C (subl.)
1g
Videos
SDS