missing translation for 'onlineSavingsMsg'
Learn More

Fullerene powder, 98+% C{70}

Product Code. 11309228
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11309228

Brand: Thermo Scientific Alfa Aesar 042601.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 115383-22-7 | C70 | 840.77 g/mol

Fullerene powder is used in chemical research. It is extensively used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115383-22-7
Molecular Formula C70
Molecular Weight (g/mol) 840.77
MDL Number MFCD00282904
InChI Key ATLMFJTZZPOKLC-UHFFFAOYSA-N
Synonym fullerene, fullerene 70, rugbyballene, carbon, 70-d5h fullerene, buckminsterfullerene, 5,6 fullerene-c70-d5h 6, c70-d5h 6 5,6 fullerene, fullerene powder, c 250mg
PubChem CID 16131935
ChEBI CHEBI:33195
IUPAC Name (C\{70}-D\{5h(6)})[5,6]fullerene
SMILES C12=C3C4=C5C6=C7C8=C9C%10=C%11C%12=C%13C%10=C%10C8=C5C1=C%10C1=C%13C5=C8C1=C2C1=C3C2=C3C%10=C%13C%14=C3C1=C8C1=C3C5=C%12C5=C8C%11=C%11C9=C7C7=C9C6=C4C2=C2C%10=C4C(=C29)C2=C6C(=C8C8=C9C6=C4C%13=C9C(=C%141)C3=C85)C%11=C27

Specifications

Color Black
Density ≈1.7 g/mL
Boiling Point 500°C to 600°C (sublimation)
Quantity 1 g
UN Number UN1325
Solubility Information Soluble in toluene and benzene.
Percent Purity ≥98%
Physical Form Powder
Assay C70
Chemical Name or Material Fullerene powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.