missing translation for 'onlineSavingsMsg'
Learn More

Fullerene, nanotube, multi-walled, as-produced cathode deposits, core and shell

Product Code. 11360128
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11360128 50 g 50g
11380128 2 g 2g
11370128 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11360128 Supplier Thermo Scientific Alfa Aesar Supplier No. 042886.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 308068-56-6 | CH4 | 16.043 g/mol

Multi-walled carbon nanotube were used in the comparative study of oxidative treatment to carbon nonmaterial's. Used as a reinforcement in CNT/SiC nano composites. Also used as fiber optics transducers, fiber optic tips for micro bubble generation in liquids, battery cathodes and high strength composites.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 308068-56-6
Molecular Formula CH4
Molecular Weight (g/mol) 16.043
MDL Number MFCD00133992
InChI Key VNWKTOKETHGBQD-UHFFFAOYSA-N
Synonym methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon
PubChem CID 297
ChEBI CHEBI:16183
IUPAC Name methane
SMILES C

Specifications

Color Black
Physical Form Powder
Quantity 50 g
UN Number UN1325
Solubility Information Insoluble in water and organic solvents.
Chemical Name or Material Fullerene, Nanotube, multi-walled, as-produced cathode deposits, core and shell

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.