Learn More
Thermo Scientific Alfa Aesar Fullerene, carbon nanotube, multi-walled, 3-20 nm OD, 1-3 nm ID, 0.1-10 micron long, 95%
Brand:  Thermo Scientific Alfa Aesar 044945.14
Fullerene, carbon nanotube, multi-walled, 3-20 nm OD, 1-3 nm ID, 0.1-10 micron long, 95%
Code : NEW
Additional Details : CAS Number : 308068-56-6 Weight : 0.02500kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III
Product Code. 15454925