Learn More
Fullerene, buckytube/nanotube, single walled, >60% SWNT, Thermo Scientific Chemicals
£107.00 - £452.00
Chemical Identifiers
CAS | 308068-56-6 |
---|---|
Molecular Formula | CH4 |
Molecular Weight (g/mol) | 16.043 |
MDL Number | MFCD00282904 |
InChI Key | VNWKTOKETHGBQD-UHFFFAOYSA-N |
Synonym | methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon |
PubChem CID | 297 |
ChEBI | CHEBI:16183 |
IUPAC Name | methane |
SMILES | C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11415220
|
Thermo Scientific Alfa Aesar
044501.77 |
0.1 g | |||||||
11405220
|
Thermo Scientific Alfa Aesar
044501.02 |
0.5 g |
£452.00
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Fullerene, buckytube/nanotube is used for several biomedical applications including the design of high-performance MRI contrast agents, X-Ray imaging contrast agents, photodynamic therapy and drug and gene delivery.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
308068-56-6 | |
16.043 | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
297 | |
methane |
CH4 | |
MFCD00282904 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
CHEBI:16183 | |
C |
Specifications
308068-56-6 | |
CH4 | |
UN1325 | |
Soluble in benzene,toluene and chloroform. | |
C | |
16.043 | |
CHEBI:16183 | |
Fullerene, Buckytube/nanotube, single walled, >60% SWNT |
Black | |
MFCD00282904 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
methane | |
297 | |
Powder |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only