Learn More
Memmert™ Full-Sight Glass Door for UF75, Max. 250°C
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF75 Oven
Unit Size:
Each
Description
- Accessory
- Full-sight
- Glass (4 layers)
- Temperature range up to 250°C
Specifications
Specifications
| For Use With (Equipment) | UF75 Oven |
| Temperature (Metric) | 250°C (Max.) |
| Type | Full-sight Door |
| Material | Glass |