Learn More
Memmert™ Full-Sight Glass Door for UF160, Max. 250°C
Product Code. 17519005 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF160 Oven
Unit Size:
Each
Product Code. 17519005
Brand: Memmert™ UF160B0
Only null left
Add to basket
Add to basket
Full-sight glass door constructed of four insulated glass layers.
  • Accessory
  • Full-sight
  • Glass (4 layers)
  • Temperature range up to 250°C
Specifications
For Use With (Equipment) UF160 Oven
Temperature (Metric) 250°C (Max.)
Type Full-sight Door
Material Glass