missing translation for 'onlineSavingsMsg'
Learn More

Full Range Indicator, pH Range 1-13, for pH Measurement, Fisher Chemical™

Product Code. 10488610 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
0.1 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10488610 500 mL 500mL
10366340 100 mL 0.1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10488610 Supplier Fisher Chemical Supplier No. U/0025/PB08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Full Range Indicator, pH Range 1-13, for pH Measurement, Fisher Chemical, Quantity: 500mL, Packaging: Plastic Coated Glass bottle, MDL Number: 283286, Physical Form: Liquid

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 283286

Specifications

Packaging Plastic Coated Glass bottle
Quantity 500 mL
Physical Form Liquid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Full range indicator, pH range 1-13
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl alcohol, Methyl alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.