missing translation for 'onlineSavingsMsg'
Learn More

Temsirolimus, MedChemExpress

Product Code. 16460598
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16460598

Brand: Medchem Express HY50910

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Temsirolimus is an inhibitor of mTOR with an IC50 of 1.76 μM. Temsirolimus activates autophagy and prevents deterioration of cardiac function in animal model.

Specifications

Chemical Name or Material Temsirolimus
Physical Form Powder
CAS 162635-04-3
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C56H87NO16
For Use With (Application) Cancer-Kinase/protease
Synonym CCI-779
Solubility Information DMSO : 250 mg/mL (242.65 mM; Need ultrasonic) ∣Ethanol : 200 mg/mL (194.12 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C([C@@]1(O)[C@@H](CC[C@@H](C[C@@H](/C(C)=C/C=C/C=C/[C@H](C[C@@H](C)C([C@@H]([C@@H](/C(C)=C/[C@H]2C)O)OC)=O)C)OC)O1)C)C(N3CCCC[C@H]3C(O[C@@H](CC2=O)[C@@H](C[C@H]4C[C@H]([C@H](OC(C(C)(CO)CO)=O)CC4)OC)C)=O)=O
Molecular Weight (g/mol) 1030.29
Formula Weight 1030.29
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.