Learn More
Fukutin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18276-1-AP-150UL
 View more versions of this product
Product Code. 16848344
  • £349.00 / 150µL
Description
FKTN regulates the migration and assembly of neurons during cortical histogenesis. Fukuyama congenital muscular dystrophy results from mutations in its gene.
Specifications
Specifications
Fukutin
Polyclonal
Unconjugated
FKTN
CMD1X, FCMD, FKTN, Fukutin, LGMD2M
Rabbit
Antigen Affinity Chromatography
RUO
2218, 246179, 362520
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75072, Q8R507
Fktn
Fukutin Fusion Protein Ag13067
150 μL
Primary
Human, Mouse, Rat