missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13541-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date.Specifications
| FUK | |
| Polyclonal | |
| Unconjugated | |
| FUK | |
| 1110046B12Rik, fucokinase, FUK, L fucose kinase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 197258, 234730, 307848 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7TMC8, Q8N0W3 | |
| FUK | |
| FUK Fusion Protein Ag4481 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction